Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfo Orange, 0.04% (w/v) Indicator Solution, Spectrum™ Chemical
SDP

Catalog No. 18614182 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-182 500 mL Poly Bottle
18-614-181 100 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-614-182 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S595500MLPL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 547-57-9

Chemical Identifiers

CAS 547-57-9 , 7732-18-5 , 1310-73-2

Specifications

Packaging Poly Bottle
Color Orange to Red
Quantity 500 mL
Grade Chemical Solution
Chemical Name or Material Sulfo Orange

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.