Learn More
MARSHALL SCIENTIFIC Crest P1200HT-45 Digital Ultrasonic Cleaner
SDP
Supplier:  MARSHALL SCIENTIFIC 1200PH0451
Encompass
The Crest P1200HT-45 Digital Ultrasonic Cleaner has a 2.5 gallon tank size and utilizes a digital heater to set temperature and total run time.
Catalog No. NC2591243