Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cresyl Diphenyl Phosphate (so called) (mixture of analogue) 93.0+%, TCI America™
SDP

Catalog No. P025925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P025925ML 25 mL
P0259500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P025925ML Supplier TCI America Supplier No. P025925ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26444-49-5
Molecular Formula C19H17O4P
MDL Number MFCD00014920
Synonym Diphenyl Cresyl Phosphate, Phosphoric Acid Cresyl Diphenyl Ester

Specifications

Color Yellow
Boiling Point 390°C
Quantity 25 mL
UN Number 3082
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material Cresyl Diphenyl Phosphate (so called) (mixture of analogue)
EINECSNumber (2)-1860&(3)-2620
RTECSNumber TC5520000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.