Learn More
StatLab CRESYL ECHT VIOLET 25GM
SDP
Supplier:  StatLab AHCEV25
Encompass_Preferred
CresylL ECHT Violet, CAS No.: 10510-54-0, Black powder; odorless, Soluble in Water, Stable, May irritate respiratory tract, May irritate skin, with itching and redness, Health: 2, Fire Hazard: 0, Reactivity: 0
Catalog No. NC9953625
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.