Description
- High brow guard for extra protection and unimpeded view
- Single lens design with integral side shields for greater orbital seal
- Spatula temple with extendable adjustment
- Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
Compliance
Spectacles meet ANSI Z87.1-1989 standards.
Specifications
Specifications
| Certifications/Compliance | Meet ANSI Z87.1-1989 safety standards |
| Lens Tint | Gray |
| Frame Color | Black |
| Lens Material | Polycarbonate |
| Frame Material | Nylon |