Learn More
MCR Safety Crews™ Tomahawk™ Safety Glasses
Catalog No. 189991909 Shop All MCR Safety Products
Change view
Click to view available options
Frame Color:
Black
Blue
Red, White, Blue
Lens Coating:
Scratch-resistant
Lens Tint:
Clear
Clear Mirror, Indoor/Outdoor
Gray
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Frame Color Lens Coating Lens Tint
18-999-1909 Red, White, Blue Scratch-resistant Clear
18-999-1905 Black Scratch-resistant Clear
19-034-304 Black - Clear
18-999-1906 Black - Gray
19-034-308 Red, White, Blue - Clear
18-999-1907 Blue Scratch-resistant Clear
19-033-449 Black - Clear Mirror, Indoor/Outdoor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 18-999-1909 Supplier MCR Safety Supplier No. TK130
Add to Cart
Add to Cart
This item is not returnable. View return policy
The Tomahawk™ is an affordable alternative for providing comfort, protection and style. Features include a stylish wrap-around design and one-piece polycarbonate lens with integral sideshields.
  • High brow guard for extra protection and unimpeded view
  • Single lens design with integral side shields for greater orbital seal
  • Spatula temple with extendable adjustment
  • Rugged polycarbonate lens filters 99.9% of harmful U.V. rays
Compliance
Spectacles meet ANSI Z87.1-1989 standards.
Specifications
Lens Coating Scratch-resistant
Certifications/Compliance Meet ANSI Z87.1-1989 safety standards
Lens Tint Clear
Frame Color Red, White, Blue
Lens Material Polycarbonate
Frame Material Nylon