Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRG-L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA14325
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA14325 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA14325 Supplier Invitrogen™ Supplier No. PA14325
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-4325 detects CRG-L2 in human samples. PA1-4325 has been successfully used in Western blotting procedures. The predicted molecular weight is 48 kDa. By Western blot, this antibody detects a protein often above ~50 kDa representing CRG-L2. This protein can form dimers. PA1-4325 immunizing peptide corresponds to amino acid residues 160-176 of human CRG-L2.

CRG-L2 is a protein that contains olfactomedin-like and collagen-like domains. The encoded protein, which exists in both transmembrane and secreted forms, promotes formation of the nodes of Ranvier in the peripheral nervous system. Mutations in this gene cause a form of lethal congenital contracture syndrome in human patients. Autoantibodies to the encoded protein have been identified in sera form patients with multifocal motor neuropathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRG-L2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Gldn
Gene Accession No. Q6ZMI3
Gene Alias cancer related gene - liver 2; cancer-related gene liver 2 protein; Clom; collomin; COLM; colmedin; Crgl2; CRG-L2; Crlg2; FLJ23917; Gldn; Gliomedin; Gliomedin shedded ectodomain; olfactomedin-related protein; UNC-112; UNQ9339/PRO34011
Gene Symbols Gldn
Host Species Rabbit
Immunogen Synthetic peptide: L(160) V G K A D E K A S E H H S P Q A(176).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342035
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.