Learn More
Stoelting™ Crile Forceps
SDP
Catalog No. 10001338 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Curved
Curved, Extra delicate
Straight
Straight, Extra delicate
Size:
5.5 in.
6.25 in.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model Size
10-001-338 Straight, Extra delicate 5.5 in.
10-001-341 Straight 5.5 in.
10-001-342 Curved 5.5 in.
10-001-340 Curved, Extra delicate 5.5 in.
10-001-343 Straight 6.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 10-001-338 Supplier Stoelting™ Supplier No. 5211020
Add to Cart
Add to Cart
Hemostatic forceps are used in many surgical procedures to control bleeding or the flow of liquids in tubing.
  • Scissor-like ring handles
  • Ratcheted handles can be locked in multiple positions in order to maintain variable levels of constant pressure
Specifications
For Use With (Application) Bleeding Control
Size 5.5 in.
Model Straight, Extra delicate