Learn More
Krackeler Scientific Crimp Caps
SDP
Catalog No. 51820552N
Change view
Click to view available options
Material (Liner):
PTFE
PTFE/Butyl
PTFE/Red Rubber
PTFE/Silicone
Closure Size:
11 mm
20 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material (Liner) Closure Size
51820552N PTFE 11 mm
056005241N PTFE/Silicone 20 mm
HP51834479N PTFE/Butyl 20 mm
HP51811210N PTFE/Red Rubber 11 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 51820552N Supplier Krackeler Scientific Supplier No. 9251820552EA
Add to Cart
Add to Cart
For headspace top vials
Specifications
Material (Liner) PTFE
Color Silver
Material Aluminum
Quantity 100 Pk.
Closure Size 11 mm
Product Type Crimp Cap