Learn More
MilliporeSigma™ Supelco™ Crimp seals with White PTFE/Silicone Septa Silver Aluminum Seal
Catalog No. 111006940 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
No. per Pack:
100
1000
Thickness (English):
2 mil
5 mil
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack Thickness (English)
11-100-6940 1000 5 mil
11-101-0165 100 2 mil
11-101-0562 100 5 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-100-6940 Supplier MilliporeSigma™ Supelco™ Supplier No. 27375
Add to Cart
Add to Cart
Crimp seals with silver aluminum seal
  • May be used to make specialized reaction vessel, by fusing them to top autosampler vials, in order to test the accuracy and reproducibility of the tetrabutylammonium fluoride and iodobenzene diacetateTBAF/PhI(OAc)2 method for detecting water in organic solvents.
  • It may also be used to seal various samples, thereby isolating them from atmospheric CO2 for a period of at least 1 month.
TRUSTED_SUSTAINABILITY
Specifications
Description Open center
Diameter (Metric) Inner 20 mm
Color White, Silver
Material PTFE, Silicone Septum, Aluminum (Seal)
No. per Pack 1000
Thickness (English) 5 mil