Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cas9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590171 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590171 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590171 Supplier Invitrogen™ Supplier No. PA590171
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: HeLa Immunogen sequence: YYVGPLARGN SRFAWMTRKS EETITPWNFE EVVDKGASAQ SFIERMTNFD KNLPNEKVLP KHSLLYEYFT VYNELTKVKY VTEGMRKPAF LSGEQKKAIV DLLFKTNRKV TVKQLKEDYF KKIECFDSVE ISGVEDRFNA SLGTYHDLLK IIKDKDFLDN EENEDILEDI VLTLTLFEDR EMIEERLKTY AHLFDDKVMK QLKRRRYTGW GRLSRKLING IRDKQSGKTI LDFLKSDGFA NRNFMQLIHD DSLTFKEDIQ KAQVSGQGDS LHEHIANLAG S.

Cas9 is an RNA-guided DNA endonuclease enzyme associated with the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) adaptive immunity system in bacteria and genetic engineering. The system is unique and flexible due to its dependence on RNA as the element that targets the nuclease to a desired DNA sequence. It can be used to induce indel mutations, specific sequence replacements/insertions, and large deletions or genomic rearrangements at any desired location in the genome. In addition, Cas9 can also be used to mediate upregulation of specific endogenous genes or to alter histone modifications or DNA methylation. Cas9 may be derived from Streptococcus pyogenes (SpCas9) or Staphylococcus aureus (SaCas9). SaCas9 has a number of properties that make it advantageous for genome editing, including its small size, high efficiency, nickase activity, and apparent specificity. SaCas9 principally recognizes an NNGRRT protospacer adjacent motif (PAM) and cleaves target DNA at rates comparable to SpCas9 with an ideal spacer lengths of 24 to 20 nucleotides. In comparison to SpCas9, SaCas9 is smaller (1368 AA versus 1053 AA) and generates indels at a comparable rate when directed against target DNA with the mutually permissive NGGRRT PAM. This makes it the preferred enzyme for all-in-one delivery of Cas9 and multiple gRNA expression cassettes with Adeno-associated vectors.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cas9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300, pH 7.3
Gene CASP9
Gene Accession No. P55211
Gene Alias cas9; CRISPR9; CRISPR-Cas9; s aureus
Gene Symbols CASP9
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 842
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.