Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ ComforTech™ Coveralls

Catalog No. 191694034 Shop All AlphaProTech Products
Change view
Click to view available options
Description:
Elastic hood, wrist with attached AquaTrak™ boots and ankle
Elastic wrist and ankle
Elastic wrist, ankle and back, attached AquaTrak™ boots
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
24 product options available for selection
Product selection table with 24 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Size
19-169-4034 Elastic wrist and ankle 6X-Large
19-165-679 Elastic wrist, ankle and back, attached AquaTrak™ boots X-Large
19-161-902B Elastic wrist and ankle Medium
19-165-678 Elastic wrist, ankle and back, attached AquaTrak™ boots Large
19-811-005 Elastic hood, wrist with attached AquaTrak™ boots and ankle 5X-Large
19-130-4225 Elastic hood, wrist with attached AquaTrak™ boots and ankle Medium
19-130-4226 Elastic hood, wrist with attached AquaTrak™ boots and ankle Large
19-161-902D Elastic wrist and ankle X-Large
19-165-677 Elastic wrist, ankle and back, attached AquaTrak™ boots Medium
19-161-902C Elastic wrist and ankle Large
19-161-902H Elastic wrist and ankle 5X-Large
19-130-4229 Elastic hood, wrist with attached AquaTrak™ boots and ankle 3X-Large
19-161-902A Elastic wrist and ankle Small
19-161-902E Elastic wrist and ankle 2X-Large
19-165-682 Elastic wrist, ankle and back, attached AquaTrak™ boots 4X-Large
19-130-4230 Elastic hood, wrist with attached AquaTrak™ boots and ankle 4X-Large
19-165-676 Elastic wrist, ankle and back, attached AquaTrak™ boots Small
19-161-902G Elastic wrist and ankle 4X-Large
19-161-902F Elastic wrist and ankle 3X-Large
19-165-681 Elastic wrist, ankle and back, attached AquaTrak™ boots 3X-Large
19-165-683 Elastic wrist, ankle and back, attached AquaTrak™ boots 5X-Large
19-130-4227 Elastic hood, wrist with attached AquaTrak™ boots and ankle X-Large
19-130-4228 Elastic hood, wrist with attached AquaTrak™ boots and ankle 2X-Large
19-165-680 Elastic wrist, ankle and back, attached AquaTrak™ boots 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 24 options available.
24 options
This item is not returnable. View return policy
Catalog No. 19-169-4034 Supplier AlphaProTech Supplier No. CVJ40229
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lightweight, durable microporous material delivers protection against particles, biologicals, and fluids in controlled environments. Available in 3 different styles.

Material rapidly dissipates vapor and heat on a microscopic level which helps to keep wearer cool and comfortable throughout the workday.

  • Material has static dissipative properties
  • Exhibits an ultra-low particle count and 99.9% filtration efficiency
  • Fluid and particulate barrier
  • Serged seams
  • Latex-free
  • White

Specifications

Product Type Cleanroom Coveralls
Size 6X-Large
Material Microporous Composite Fabric
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Closure Type Zipper
Seam Type Serged
Sterility Non-sterile
Description Elastic wrist and ankle
Latex Free Yes
For Use With (Application) For use in controlled environments and barrier to particles, biologicals, and fluids with 99.9% filtration efficiency
Style ComforTech™
Cleanroom Class Class 10-100,000 (ISO 4-8)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.