Description
- Lightweight and easily fits over glasses
- Double-sided anti-fog coating
- Low distorting and 100% optically clear
- Soft polyurethane foam headband fits snugly against the forehead
- Full-face style (Cat. Nos. 23-309-829 and 23-309-828) provides complete coverage including the sides of the face
Specifications
Specifications
| Lens Coating | Anti-fog |
| Thickness (English) | 7 mil shield |
| Description | Fits easily over glasses |
| Lens Tint | Clear |
| Headband Material | Adjustable Hook-and-Loop-Closure |
| No. per Case | 100/Cs. |
| Model | Half-face |