Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ Microbreathe™ Coveralls

Catalog No. 19811660 Shop All AlphaProTech Products
Change view
Click to view available options
Size:
Large
3X-Large
5X-Large
6X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-811-660 6X-Large
19-811-657 3X-Large
19-811-659 5X-Large
19-811-655 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-811-660 Supplier AlphaProTech Supplier No. CV046329TH
This item is not returnable. View return policy

Strong, flexible Microbreathe fabric coveralls provide outstanding protection, breathability, and superior barrier properties in controlled environments.

  • Barrier to particles, biologicals and fluids
  • Excellent vapor transmission rate (breatheability)
  • High filtration efficiency with low particle levels
  • Static dissipative

Specifications

Product Type Cleanroom Coveralls
Size 6X-Large
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic, Thumb Loop
Closure Type Zipper
Seam Type Bound
Sterility Non-sterile
Back Style Elastic
Description Elastic wrist, ankle and back, Thumb loop
For Use With (Application) For use in controlled environments and barrier to particles, biologicals, and fluids with 99.9% filtration efficiency
Style MicroBreathe™
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.