Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AlphaProTech Critical Cover™ PFL™ Face Masks with Magic Arch™
BIOPHARMA

ASTM Level 3 face masks feature a Positive Facial Lock™ (PFL) utilizes Twist-Seal™ technology to create a custom fit and significantly reduces the possibility of inhaling airborne contaminants.

Supplier:  AlphaProTech 615PD

 View more versions of this product

Catalog No. 19-168-217

This item is not returnable. View return policy

Description

Description

  • Medical face masks used in health care services such as surgery and patient care
  • Positive Facial Lock™ (PFL™) feature utilizes Twist-Seal technology that allows a custom fit
  • Twist-Seal technology significantly reduces the possibility of the user inhaling airborne contaminants
  • Seal fits a variety of facial sizes and features
  • Magic Arch™ prevents the 8” mask from coming in contact with nose and mouth making it easier to breathe
  • PFL masks are fluid resistant and create a durable exterior and a soft, comfortable interior
  • Two colors in headband or ear loop style. FCAT #19-166-100 is packaged in a cleanroom environment
  • ASTM F2100 Compliant Level 3 mask
Specifications

Specifications

Medical/Procedural Face Mask
Purple
Standard
Pleated
≥98% (PFE), ≥98% (BFE)
615PD
Dispenser box
Fluid
ASTM Level 3
Earloops
Non-sterile
Single-use
Surgery, patient care, healthcare services
300/Cs.
0.1 μm
Critical Cover™ PFL™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.