Learn More
Invitrogen™ Crk Monoclonal Antibody (M332)

Description
This antibody detects a 40 kDa* protein corresponding to the molecular mass of Crk II on SDS-PAGE immunoblots of rat PC12 and human Jurkat cells.
Specifications
Specifications
| Antigen | Crk |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | M332 |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide |
| Gene | Crk |
| Gene Accession No. | P46108, P47811, P70618, Q16539, Q63768, Q64010 |
| Gene Alias | Adapter molecule crk; avian sarcoma virus CT10 (v-crk) oncogene homolog; c-Crk; CRK; CRK proto-oncogene, adaptor protein; CRK1; Crk3; Crk-I; CRKII; Crk-II; CrkIII; Crk-III; Crko; CSAID-binding protein; Csaids binding protein; CSBP; Csbp1; CSBP2; CSPB1; CT-10 related kinase 3; cytokine suppressive anti-inflammatory drug binding protein; cytokine suppressive anti-inflammatory drug binding protein 1; cytokine suppressive anti-inflammatory drug-binding protein; cytokine-supressive anti-inflammatory drug binding protein; Exip; Hog; I79_016605; MAP kinase 14; MAP kinase 2; MAP kinase Mxi2; MAP kinase p38 alpha; MAPK 14; MAPK14; MAX-interacting protein 2; mitogen activated protein kinase 14; mitogen-activated protein kinase 14; mitogen-activated protein kinase 14A; mitogen-activated protein kinase p38 alpha; MXI2; p38; p38 alpha; p38 MAP kinase; p38 MAP kinase alpha; p38 MAPK; p38 mitogen activated protein kinase; p38a; p38ALPHA; p38-alpha; p38alpha Exip; p38Hog; p38MAPK; PRKM14; PRKM15; Proto-oncogene c-Crk; reactive kinase; RK; SAPK2A; stress-activated protein kinase 2A; tRNA synthetase cofactor p38; v-crk avian sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene homolog; v-crk sarcoma virus CT10 oncogene homolog (avian); v-crk sarcoma virus CT10 oncogene-like protein |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.