Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRLS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100137
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100137 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100137 Supplier Invitrogen™ Supplier No. PA5100137
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total CRLS1.

The function of this protein is still unknown. The gene of human hypothetical protein (LOC54675) is up-regulated in colon cancer metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRLS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene CRLS1
Gene Accession No. Q5U2V5, Q80ZM8, Q9UJA2
Gene Alias 0610009I22Rik; 4930557M15Rik; 5730490M08Rik; C20orf155; cardiolipin synthase; cardiolipin synthase (CMP-forming); cardiolipin synthase 1; cardiolipin synthetase; CLS; CLS1; CRLS1; dJ967N21.6; GCD10; Protein GCD10 homolog; RGD1311037
Gene Symbols CRLS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human CRLS1(Accession Q9UJA2), corresponding to amino acid residues M197-A247.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 366196, 54675, 66586
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.