Learn More
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: rat liver tissue, rat lung tissue, mouse liver tissue, mouse lung tissue, human HepG2 whole cell, human SMMC-7721 whole cell, human Hela whole cell, human Jurkat whole cell. IHC: human lung cancer tissue, human intestinal cancer tissue. ICC/IF: U20S cell. Flow: SiHa cell.
Specifications
Specifications
| Antigen | CRM1 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | 5G3 |
| Concentration | 500 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 4mg trehalose and 0.05mg sodium azide |
| Gene | XPO1 |
| Gene Accession No. | O14980, Q6P5F9, Q80U96 |
| Gene Alias | AA420417; chromosome region maintenance 1 homolog; chromosome region maintenance 1 protein homolog; CRM1; emb; Exp1; exportin 1; exportin 1 (CRM1 homolog, yeast); exportin 1 (CRM1, yeast, homolog); exportin 1, CRM1 homolog; exportin 1, CRM1 homolog (yeast); exportin-1; exportin-1 (required for chromosome region maintenance); Xpo1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
