Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587417 Supplier Invitrogen™ Supplier No. PA587417
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, B-cell, Mouse liver; Cellular Location: Cajal body, Cytoplasm, Nucleus, nucleolus, nucleoplasm Immunogen sequence: PGNPVNNQIF LQEYVANLLK SAFPHLQDAQ VKLFVTGLFS LNQDIPAFKE HLRDFLVQIK EFAGEDTSDL FLEEREIALR QADEEKHKRQ MSVPGIFNPH E.

Exportin - 1 (XPO1) mediates the nuclear export of cellular proteins bearing a leucine-rich nuclear export signal (NES) and RNAs. In the nucleus, in association with RANBP3, XPO1 binds cooperatively to the NES on its target protein and to the GTPase RAN in its active GTP-bound form (Ran-GTP). Docking of this complex to the nuclear pore complex is mediated through binding to nucleoproteins. Upon transit of a nuclear complex into the cytoplasm, disassembling of the complex and hydrolysis of Ran-GTP to Ran-GDP cause release of the cargo from the export receptor. The directionality of the nuclear export is thought to be conferred by an asymmetric distribution of the GTP and GDP bound forms of Ran between the cytoplasm and nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRM1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene XPO1
Gene Accession No. O14980, Q6P5F9
Gene Alias AA420417; chromosome region maintenance 1 homolog; chromosome region maintenance 1 protein homolog; CRM1; emb; Exp1; exportin 1; exportin 1 (CRM1 homolog, yeast); exportin 1 (CRM1, yeast, homolog); exportin 1, CRM1 homolog; exportin 1, CRM1 homolog (yeast); exportin-1; exportin-1 (required for chromosome region maintenance); Xpo1
Gene Symbols XPO1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 964-1064 of human CRM1/CRM1/XPO1 (NP_0033911).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103573, 7514
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.