Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Crocein Scarlet 7B, >70% dye content

Catalog No. AAJ6687606
Change view
Click to view available options
Quantity:
1g
5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6687606 5g
AAJ6687603 1g
AAJ6687614 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6687606 Supplier Thermo Scientific Chemicals Supplier No. J6687606
Add to Cart

CAS: 6226-76-2 | C24H18N4Na2O7S2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6226-76-2
Molecular Formula C24H18N4Na2O7S2
Synonym Acid Red 71; C.I. 27165

Specifications

Color Red
Form Powder
Format Powder
Formula Weight 584.53
Melting Point 181-188°
MDL Number MFCD00056588
Purity >70% dye content
Quantity 5g
InChI Key GMWXFMFHCCFVJU-NPYGFOBASA-L
SMILES [Na+].[Na+].CC1=C(C=CC(=C1)\N=N\C1=C(C)C=C(C=C1)S([O-])(=O)=O)\N=N\C1=C2C(C=CC=C2S([O-])(=O)=O)=CC=C1O
IUPAC Name disodium 7-hydroxy-8-[(1E)-2-{2-methyl-4-[(1E)-2-(2-methyl-4-sulfonatophenyl)diazen-1-yl]phenyl}diazen-1-yl]naphthalene-1-sulfonate
Molecular Weight (g/mol) 584.53
Solubility Soluble in water
Product Type Crocein Scarlet 7B
Show More Show Less
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.