Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Croconic acid disodium salt, 97%

Catalog No. aab2223501
Change view
Click to view available options
Quantity:
0.25 g
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2223501 0.25 g
AAB2223503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2223501 Supplier Thermo Scientific Chemicals Supplier No. B2223501
Only null left
Add to Cart

CAS: 14379-00-1 | C5Na2O5 | 186.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14379-00-1
Molecular Formula C5Na2O5
Molecular Weight (g/mol) 186.03
MDL Number MFCD00191954
InChI Key OQXLFPHHAAAVKQ-UHFFFAOYSA-L
Synonym croconic acid disodium salt, croconic acid, disodium salt, disodium trioxocyclopent-1-ene-1,2-bis olate, sodium 3,4,5-trioxocyclopent-1-ene-1,2-bis olate, 4,5-dihydroxy-4-cyclopentene-1,2,3-trione disodium salt
PubChem CID 12120285
IUPAC Name disodium;3,4,5-trioxocyclopentene-1,2-diolate
SMILES [Na+].[Na+].[O-]C1=C([O-])C(=O)C(=O)C1=O

Specifications

Melting Point >300°C
Quantity 0.25 g
Formula Weight 186.03
Percent Purity 97%
Chemical Name or Material Croconic acid disodium salt
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.