Learn More
Apexbio Technology LLC Cromolyn sodium 15826-37-6 5g
SDP
Supplier:  Apexbio Technology LLC B33005000
Encompass_Preferred
Cromolyn sodium (CAS 15826-37-6) is a small molecule that functions as a mast cell membrane stabilizer It inhibits antigen-induced histamine release by preventing degranulation of sensitized mast cells Due to minimal gastrointestinal absorption cromolyn sodium is typically administered via inhalation in powder form Preclinical studies have demonstrated that cromolyn sodium suppresses both immediate and late-phase asthmatic responses and reduces the release of non-histamine chemical mediators Additionally it prevents ciliary transport impairment associated with bronchial provocation and can mitigate symptoms in exercise-induced asthma models These properties make cromolyn sodium a useful tool for investigating allergic and inflammatory mechanisms in respiratory research
Catalog No. 50-000-96483
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.