Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cromolyn Sodium, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18601875 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-875 25 g Amber Glass Bottle
18-601-876 100 g Amber Glass Bottle
18-601-877 500 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-601-875 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C136025GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

C1360, 15826-37-6, C23H14Na2O11

Spectrum™ Chemical Cromolyn Sodium, USP is an anti-inflammatory medication. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 15826-37-6
Molecular Formula C23H14Na2O11
Molecular Weight (g/mol) 512.33
MDL Number MFCD00057744
InChI Key VLARUOGDXDTHEH-UHFFFAOYSA-L
IUPAC Name disodium 5-{3-[(2-carboxylato-4-oxo-4H-chromen-5-yl)oxy]-2-hydroxypropoxy}-4-oxo-4H-chromene-2-carboxylate
SMILES [Na+].[Na+].OC(COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O)COC1=C2C(=O)C=C(OC2=CC=C1)C([O-])=O

Specifications

CAS Max % 100%
Quantity 25 g
Percent Purity 98 to 101%
Grade USP
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.