Learn More
ATCC Pseudomonas syringae; pathovar tomato; genomic DNA
Supplier:  ATCC BAA871D5
Encompass
Genomic DNA isolated from Pseudomonas syringae patovar tomato strain DC3000. This bacterial strain is also available as ATCC Catalog No. BAA-871™.
Catalog No. 50-238-1016
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.