Learn More
CROP/Luc7A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Bovine and Orangutan
Specifications
Specifications
| Antigen | CROP/Luc7A |
| Applications | Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | 1 mg/ml |
| Conjugate | Unconjugated |
| Formulation | phosphate, tris citrate with 0.09% sodium azide; pH 7-8 |
| Gene | LUC7L3 |
| Gene Accession No. | O95232 |
| Gene Alias | cAMP regulatory element-associated protein 1, cisplatin resistance associated overexpressed protein, cisplatin resistance-associated overexpressed protein, cisplatin resistance-associated-overexpressed protein, CRA, CREAP-1, CRE-associated protein, CRE-associated protein 1, CROP, FLJ11063, hLuc7A, LUC7A, LUC7-like 3 (S. cerevisiae), LUC7-like 3 pre-mRNA splicing factor, OA48-18, okadaic acid-inducible phosphoprotein OA48-18 |
| Gene Symbols | LUC7L3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.