Learn More
CROP/Luc7A Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301499AT
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse, Bovine and Orangutan
The function of this protein remains unknown.Specifications
| CROP/Luc7A | |
| Polyclonal | |
| Unconjugated | |
| LUC7L3 | |
| cAMP regulatory element-associated protein 1, cisplatin resistance associated overexpressed protein, cisplatin resistance-associated overexpressed protein, cisplatin resistance-associated-overexpressed protein, CRA, CREAP-1, CRE-associated protein, CRE-associated protein 1, CROP, FLJ11063, hLuc7A, LUC7A, LUC7-like 3 (S. cerevisiae), LUC7-like 3 pre-mRNA splicing factor, OA48-18, okadaic acid-inducible phosphoprotein OA48-18 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51747 | |
| 4° C | |
| Liquid |
| Immunoprecipitation | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| O95232 | |
| LUC7L3 | |
| Between 200 and 250 | |
| 10 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.