Learn More
IDEX Health & Science Upchurch CROSS HP PEEK .050 THRU HOLE
Supplier:  IDEX Health & Science Upchurch P726
Encompass_Preferred
Idex High-Pressure Crosses, Pressure Rating: 5000 psi, Diameter Inner: 0.05 in., Diameter Outer: 1.25 mm, For Use With: 1/16 in. O.D. Tubing, PEEK
Catalog No. NC1919106
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.