Learn More
Enzo Life Sciences Crosstide (1mg). CAS: 171783-05-4
SDP
Supplier:  Enzo Life Sciences BMLP1490001
Encompass_Preferred
This peptide sequence, GRPRTSSFAEG, corresponds to the sequence of GSK3 surrounding the serine phosphorylated by MAP kinase activated protein kinase (MAPKAP kinase-1) and p70 ribosomal S6 kinase. Acts as a substrate for Akt (PKB), Km=4µM. Useful in phosphocellulose kinase assays. Long Term Storage: -20°C.
Catalog No. 50-201-1257
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.