Learn More
Apexbio Technology LLC Crotaline 315-22-0 100mg
SDP
Supplier:  Apexbio Technology LLC N1470100
Encompass_Preferred
Crotaline is a pyrrolizidine alkaloid derived from the plant Crotalaria assamica displaying antitumor properties of interest to biomedical research This naturally occurring alkaloid consists structurally of a pyrrolizidine ring framework which can interact with biological macromolecules influencing cellular processes relevant to cancer proliferation and apoptosis pathways It is routinely employed in experimental work focusing on investigating molecular mechanisms underlying tumorigenesis and exploring potential therapeutic strategies against cancers For in vitro research applications crotaline is compatible with solvents including methanol ethanol and chloroform and should be stored under tightly sealed cool and dry conditions to maintain chemical stability and experimental reliability
Catalog No. 50-000-62822
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.