Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Croton Oil, Spectrum™ Chemical
SDP

Catalog No. 18601884 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
1 kg
12 kg
Packaging:
Amber Glass Bottle
Steel Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-884 25 g Amber Glass Bottle
18-601-883 10 g Amber Glass Bottle
18-601-885 100 g Amber Glass Bottle
18-601-886 500 g Amber Glass Bottle
18-601-887 1 kg Amber Glass Bottle
18-601-888 12 kg Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-601-884 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CR13225GM
Add to Cart
Add to Cart

CR132, 8001-28-3

Spectrum™ Chemical Croton Oil is an oil prepared from the seeds of the tree, Croton tiglium. Croton oil is the source of the organic compound phorbol and is the basis of rejuvenating chemical peels, due to the caustic exfoliating effects it has on the dermal components of the skin. Used in conjunction with phenol solutions, it results in an intense reaction which leads to initial skin sloughing and then eventual regeneration. In traditional Chinese medicine it is used as an ingredient in some liniments.

Chemical Identifiers

CAS 8001-28-3

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Refractive Index 1.4760-1.4780
Quantity 25 g
UN Number UN2810
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.