Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotonyl chloride, tech. 90%

Catalog No. AAA1371022
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1371022 100 g
AAA1371014 25 g
AAA1371036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1371022 Supplier Thermo Scientific Chemicals Supplier No. A1371022
Add to Cart

CAS: 625-35-4 | C4H5ClO | 104.533 g/mol

Crotonyl chloride is used in agrochemical, pharmaceutical and dyestuff field. Crotonoyl chloride was used in the synthesis of α,β-unsaturated ester.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Crotonyl chloride is used in agrochemical, pharmaceutical and dyestuff field. Crotonoyl chloride was used in the synthesis of α,β-unsaturated ester.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Store at 4°C. Store away from heat and moisture. Incompatible with
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 625-35-4
Molecular Formula C4H5ClO
Molecular Weight (g/mol) 104.533
MDL Number MFCD00000734
InChI Key RJUIDDKTATZJFE-NSCUHMNNSA-N
Synonym crotonyl chloride, 2-butenoyl chloride, crotonoyl chloride, e-but-2-enoyl chloride, e-2-butenoyl chloride, crotonic acid chloride, 2e-but-2-enoyl chloride, 2-butenoyl chloride, e, 2-butenoyl chloride, 2e, trans-2-butenoyl chloride
PubChem CID 643131
IUPAC Name (E)-but-2-enoyl chloride
SMILES CC=CC(=O)Cl

Specifications

Density 1.091
Boiling Point 121°C to 123°C
Flash Point 35°C (95°F)
Odor Acrid
Refractive Index 1.46
Quantity 100 g
UN Number UN2920
Beilstein 506118
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 104.54
Percent Purity ≥90%
Assay remainder mainly cis-isomer
Chemical Name or Material Crotonyl chloride
Show More Show Less
Hazard Category H226-H314
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber 210-889-9
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.