Learn More
METTLER TOLEDO Crucible 100 μL Glass
Catalog No. 01912700 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
100 μL
Catalog No. Capacity (Metric)
01-912-700 100 μL
1 options
Catalog No. 01-912-700 Supplier METTLER TOLEDO Supplier No. 27812
Set of 50 crucibles. Duran glass crucibles are transparent and chemically resistant. The crucible is sealed by melting the neck in a small flame. These crucibles are only suitable for DSCs with furnace expander. Crucible presses sold separately.
  • Duran glass crucibles designed for specialized DSC applications
  • 100 μL capacity for accurate thermal analysis of larger samples
  • Supports up to 5 MPa pressure for reliable performance
  • Endures temperatures up to 500°C
  • No lids, no-pin design; supplied in sets of 50 crucibles
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Capacity (Metric) 100 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder