Learn More
METTLER TOLEDO Crucible 160 μL Al
Catalog No. 01912699 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
160 μL
Catalog No. Capacity (Metric)
01-912-699 160 μL
1 options
Catalog No. 01-912-699 Supplier METTLER TOLEDO Supplier No. 27811
Add to Cart
Add to Cart
Set of 40 aluminum crucibles with lids, designed for DSC measurements of samples showing very weak effects. Due to their height and larger volume, temperature gradients within the sample may occur. Crucible presses are available separately.
  • Aluminum crucibles for DSC analysis
  • 160 μL volume with lids and pin
  • Withstands up to 640°C
  • Maximum pressure of 0.2 MPa
  • Supplied in sets of 40 pieces
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Capacity (Metric) 160 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder