Learn More
METTLER TOLEDO Crucible 25 μL Al
Catalog No. 01912701 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
25 μL
Catalog No. Capacity (Metric)
01-912-701 25 μL
1 options
Catalog No. 01-912-701 Supplier METTLER TOLEDO Supplier No. 30085850
Add to Cart
Add to Cart
Set of 100 aluminum crucibles with lids, optimized for enthalpy measurements. Designed for DSC analysis of volatile substances, they can be hermetically sealed to minimize evaporation or vaporization. Crucible presses sold separately.
  • Aluminum crucibles for DSC enthalpy measurements
  • 25 μL volume with included lids
  • Withstands up to 640°C
  • Maximum pressure of 0.2 MPa
  • Supplied in sets of 100 pieces
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Capacity (Metric) 25 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder