Learn More
METTLER TOLEDO Crucible 30 μL Pt
Catalog No. 01912698 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
30 μL
Catalog No. Capacity (Metric)
01-912-698 30 μL
1 options
Catalog No. 01-912-698 Supplier METTLER TOLEDO Supplier No. 51140842
Add to Cart
Add to Cart
Set of 4 crucibles with lids. Platinum crucibles are mainly used for TGA or DSC measurements at temperatures above 640 °C. SDTA and DSC curves measured with platinum crucibles are usually better than those obtained using crucibles made of alumina. Crucible presses sold separately.
  • Platinum crucibles engineered for precise high-temperature thermal analysis
  • 30 μL volume capacity for accurate and reproducible measurements
  • Withstands extreme conditions up to 1600°C
  • No-pin design supplied with secure lids
  • Durable platinum construction; supplied in packs of 4 crucibles
TRUSTED_SUSTAINABILITY
Specifications
Material Platinum
Capacity (Metric) 30 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder