Learn More
METTLER TOLEDO Crucible 40 μL Al
Catalog No. 01912687 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
40 μL
Catalog No. Capacity (Metric)
01-912-687 40 μL
1 options
Catalog No. 01-912-687 Supplier METTLER TOLEDO Supplier No. 27331
Add to Cart
Add to Cart
Set of 100 aluminum crucibles with lids, designed for precise DSC analysis. The shallow shape with a flat base ensures minimal temperature gradients for accurate measurements. Crucible presses available separately.
  • Aluminum crucibles for reliable DSC measurements
  • 40 μL volume with included lids and pin
  • Withstands up to 640°C
  • Maximum pressure of 0.2 MPa
  • Supplied in sets of 100 pieces
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Capacity (Metric) 40 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder