Learn More
METTLER TOLEDO Crucible 40 μL Al Gold Plated
Catalog No. 01674714 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
40 μL
Catalog No. Capacity (Metric)
01-674-714 40 μL
1 options
Catalog No. 01-674-714 Supplier METTLER TOLEDO Supplier No. 51142973
Add to Cart
Add to Cart
Set of 10 gold-plated aluminum crucibles with lids. Chemically resistant and suitable for hermetic sealing by cold welding. Maximum permissible pressure is 0.2 MPa. Crucible presses sold separately.
  • Designed for demanding DSC applications requiring chemical resistance
  • 40 μL capacity for precise sample analysis under controlled conditions
  • Withstands temperatures up to 350°C and a maximum pressure of 0.2 MPa
  • Hermetically sealable through cold welding for secure measurements
  • Supplied as a set of 10 crucibles with lids for reliable results
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Capacity (Metric) 40 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder