Learn More
METTLER TOLEDO Crucible 40 μL Au
Catalog No. 01912709 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
40 μL
Catalog No. Capacity (Metric)
01-912-709 40 μL
1 options
Catalog No. 01-912-709 Supplier METTLER TOLEDO Supplier No. 27220
Add to Cart
Add to Cart
Set of 6 gold crucibles with lids. Chemically resistant and suitable for hermetic sealing by cold welding. Maximum permissible pressure is 0.25 MPa. Crucible presses sold separately.
  • Gold crucibles designed for chemically resistant DSC applications
  • 40 μL volume capacity for precise thermal analysis
  • Withstands temperatures up to 750°C for reliable results
  • No-pin design ensures accurate sealing and handling
  • Supplied in sets of 6 crucibles with lids for efficient use
TRUSTED_SUSTAINABILITY
Specifications
Material Gold
Capacity (Metric) 40 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder