Learn More
METTLER TOLEDO Crucible 70 μL PCA
Catalog No. 01912693 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
70 μL
Catalog No. Capacity (Metric)
01-912-693 70 μL
1 options
Catalog No. 01-912-693 Supplier METTLER TOLEDO Supplier No. 51140845
Add to Cart
Add to Cart
Set of 4 crucibles with lids made of polycrystalline alumina oxide (PCA) with properties similar to sapphire. Chemically resistant, inert, and recommended for all materials, including melting metals. Crucible presses sold separately.
  • PCA crucibles with sapphire-like properties for advanced DSC applications
  • 70 μL capacity suitable for a wide range of materials
  • Withstands temperatures up to 2000°C for extreme conditions
  • Chemically resistant and inert for reliable thermal analysis
  • No-pin design with lids included; supplied in sets of 4 pieces
TRUSTED_SUSTAINABILITY
Specifications
Material PCA/Sapphire
Capacity (Metric) 70 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder