Learn More
METTLER TOLEDO Crucible HP 30 μL SST/Au
Catalog No. 01912706 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
30 μL
Catalog No. Capacity (Metric)
01-912-706 30 μL
1 options
Catalog No. 01-912-706 Supplier METTLER TOLEDO Supplier No. 51140405
Add to Cart
Add to Cart
Set of 3 gold-plated crucibles with screw caps and seals. This flat crucible delivers improved DSC signals compared to larger high-pressure types. Reusable up to 20 times after cleaning. Crucible presses sold separately.
  • High-pressure steel/Au crucibles with gold-plated surface for precise DSC thermal analysis
  • 30 μL capacity with flat, lightweight design delivering stronger DSC signals
  • Withstands up to 15 MPa pressure and temperatures up to 350°C
  • Includes screw caps and seals; no-pin design for secure measurements
  • Reusable up to 20 times after cleaning; supplied in packs of 3 crucibles
TRUSTED_SUSTAINABILITY
Specifications
Material Steel
Capacity (Metric) 30 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder