Learn More
METTLER TOLEDO Crucible HP 30 μL SST
Catalog No. 01912695 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
30 μL
Catalog No. Capacity (Metric)
01-912-695 30 μL
1 options
Catalog No. 01-912-695 Supplier METTLER TOLEDO Supplier No. 51140404
Add to Cart
Add to Cart
Set of 3 crucibles with screw caps and seals. This flat crucible delivers stronger DSC signals than larger high-pressure crucibles. Reusable up to 20 times after cleaning. Crucible presses sold separately.
  • Stainless steel crucibles designed for high-pressure DSC applications
  • 30 μL capacity with flat, lightweight design for accurate signal detection
  • Withstands up to 15 MPa pressure for reliable thermal analysis
  • Endures extreme conditions up to 750 °C without compromising results
  • No-pin configuration with secure sealing for precise laboratory use
TRUSTED_SUSTAINABILITY
Specifications
Material Steel
Capacity (Metric) 30 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder