Description
- Gold-plated stainless steel crucibles for reliable performance
- 40 μL volume for precise sample testing
- Withstands up to 15 MPa pressure and 400°CC temperature
- Includes lids and rupture disks, supplied without pins
- Pack of 200 single-use crucibles
Specifications
Specifications
| Material | Steel |
| Capacity (Metric) | 40 μL |
| Autoclavable | Not Autoclavable |
| Product Type | Crucible |
| Shape | Cylinder |