Learn More
METTLER TOLEDO Crucible HP 40 μL SST/Au
Catalog No. 01912696 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
40 μL
Catalog No. Capacity (Metric)
01-912-696 40 μL
1 options
Catalog No. 01-912-696 Supplier METTLER TOLEDO Supplier No. 26732
Add to Cart
Add to Cart
Set of 25 crucibles (with pin), lids and rupture disks. The gold-plated steel crucibles have proven highly effective in the field of safety investigations. They can only be used for one measurement each. The maximum pressure is 15 MPa. Crucible presses sold separately.
  • Gold-plated stainless steel crucibles ensure consistent and reliable results
  • 25 μL sample volume suitable for safety and thermal analysis
  • Rated for up to 15 MPa maximum pressure and 400°CC operation
  • Includes lids, supplied with pins
  • Pack of 25 crucibles for single-use applications
TRUSTED_SUSTAINABILITY
Specifications
Material Steel
Capacity (Metric) 40 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder