Learn More
METTLER TOLEDO Crucible MP 120 μL SST, Pin
Catalog No. 01912708 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
120 μL
Catalog No. Capacity (Metric)
01-912-708 120 μL
1 options
Catalog No. 01-912-708 Supplier METTLER TOLEDO Supplier No. 26929
Add to Cart
Add to Cart
Set of 25 crucibles, lids, and seals. This medium-pressure crucible is sealed with an FPM O-ring, withstanding pressures up to 2 MPa. It can also be used without an O-ring to generate a self-contained atmosphere. Crucible presses sold separately.
  • Durable stainless steel crucibles for medium-pressure DSC applications
  • 120 μL capacity with lids and sealing pin for secure handling
  • Withstands up to 2 MPa pressure and temperatures up to 250°C
  • Supplied in sets of 25 pieces with FPM O-ring sealing option
  • Can also operate without O-ring for self-generated atmosphere
TRUSTED_SUSTAINABILITY
Specifications
Material Steel
Capacity (Metric) 120 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder