Learn More
METTLER TOLEDO Crucible MP 120 μL SST
Catalog No. 01912705 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
120 μL
Catalog No. Capacity (Metric)
01-912-705 120 μL
1 options
Catalog No. 01-912-705 Supplier METTLER TOLEDO Supplier No. 29990
Add to Cart
Add to Cart
Set of 25 crucibles with lids and seals for medium-pressure applications up to 2 MPa, sealed with an FPM O-ring or without for self-generated atmosphere. Crucible presses sold separately.
  • Durable stainless steel/Au crucibles for medium-pressure DSC applications
  • 120 μL capacity with lids and sealing pin for secure handling
  • Withstands up to 2 MPa pressure and temperatures up to 250°C
  • Supplied in sets of 25 pieces with FPM O-ring sealing option
  • Can also operate without O-ring for self-generated atmosphere
TRUSTED_SUSTAINABILITY
Specifications
Material Steel
Capacity (Metric) 120 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder