Learn More
METTLER TOLEDO Crucible Pt
Catalog No. 01912707 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Capacity (Metric):
70 μL
150 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
01-912-707 150 μL
01-912-694 70 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-912-707 Supplier METTLER TOLEDO Supplier No. 24126
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Set of 4 crucibles with lids. Platinum crucibles are mainly used for TGA or DSC measurements at temperatures above 640 °C.
  • Includes lids for secure sample containment
  • Provides superior SDTA and DSC curves compared to alumina crucibles
  • Compatible with high-temperature analytical instruments
  • Crucible presses sold separately
  • Set of 4 crucibles for efficient sample processing
TRUSTED_SUSTAINABILITY
Specifications
Material Platinum
Capacity (Metric) 150 μL
Autoclavable Not Autoclavable
Product Type Crucible
Shape Cylinder