Learn More
Eisco™ Crucible Tongs with Bow
SCI_ED
Catalog No. S27100 Shop All Eisco Products
Change view
Click to view available options
Length (Metric):
24 cm
Catalog No. Length (Metric)
S27100 24 cm
1 options
Catalog No. S27100 Supplier Eisco™ Supplier No. CH0729B
Add to Cart
Edge
Add to Cart
General purpose tongs with small bow and straight, serrated tips designed for grasping hot crucibles, evaporating dishes, or small flasks
  • General purpose
  • Metal crucible tongs
  • Small bow with straight, serrated tips
  • Designed for grasping hot crucibles, evaporating dishes, or small flasks
Specifications
Product Type Crucible Tongs
Length (Metric) 24 cm
Tip Style Straight, Serrated
Material Metal