Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kodiak™ Crusade CT WP 6 in. Black Boot

Offers a full-grain waterproof leather upper, waterproof membrane, an ASTM approved composite toe, slip and oil-resistant rubber outsole, and electrical hazard protection to help protect your feet, and confidently tackle tough days or terrain on the job-site.

Supplier:  Kodiak™ K4NKBK 100 W

 View more versions of this product

Catalog No. 17-100-250


Only null left
Add to Cart

Description

Description

  • Full-grain waterproof leather upper with waterproof membrane construction
  • KODIAK™ SOS Smell Out Science™ odor-control technology
  • Abrasion-resistant leather toe guard offers added durability
  • Full leather bellows tongue
  • Removable, comfort trainer perforated EVA insole
  • Kodiak™ Ultimate SR and oil-resistant rubber outsole with EVA midsole offers maximum comfort
Specifications

Specifications

Black
10W
Yes
Boot
Leather
Men's
Yes
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.