Learn More
Description
PA1-527 detects recombinant human cryptochrome 1 (Cry1). PA1-527 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~70 kDa protein representing full length recombinant human Cry1 protein. PA1-527 immunizing peptide corresponds to amino acid residues 594-606 from mouse Cry1. PA1-527 immunizing peptide (Cat. # PEP-103) is available for use in neutralization and control procedures.
Specifications
Specifications
| Antigen | CRY1 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | CRY1 |
| Gene Accession No. | P97784, Q16526 |
| Gene Alias | AU020726; AU021000; Cry1; cryptochrome 1 (photolyase-like); cryptochrome circadian clock 1; cryptochrome-1; PHLL1 |
| Gene Symbols | CRY1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
