Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CRYAB Monoclonal Antibody (3A10.C9), APC
GREENER_CHOICE

Catalog No. PIMA545217
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA545217 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA545217 Supplier Invitrogen™ Supplier No. MA545217
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

0.5 μg/mL of MA5-45217 was sufficient for detection of 50 ng purified alpha B crystalline by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 20kDa (predicted mol.weight is approximately 21kDa). Does not cross-react with alpha A-crystallin, beta L-crystallin, beta H-crystallin, gamma -crystallin, HSP25, HSP27 or HSP47 proteins.

Lens proteins consist almost entirely of crystallins (about 95%). Crystallins are also found vertebrate skeletal muscle tissue. In the lens, their structural function is to assist in maintaining the proper refractive index of the lens. The mammalian lens contains 3 major classes of crystallins: alpha, beta, and gamma. Alpha-crystallin is the largest of the crystallins and is composed of 2 primary gene products-alpha-A and alpha-B. There are at least 5 different proteins comprising the beta-crystallins. The gamma-crystallins are monomeric, but there are at least 5 gamma crystallins identified in bovine and rat lens. Alpha-Crystallin comprises 40% of total lens protein composition. In addition to maintaining proper refractive index, it also functions in a chaperone like manner by preventing the formation of aggregates possibly leading to cataract formation. It is believed that the phosphorylated states of the alpha-crystallin occur in response to cellular stress and may serve a structural control function and play a role in protein maintenance. Alpha-B crystallin has been linked to Alexander's disease where it accumulates in brain cells of those afflicted.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CRYAB
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3A10.C9
Concentration 1 mg/mL
Conjugate APC
Formulation 2.48mM MES, 95.46mM phosphate with 2mM EDTA and no preservative; pH 7.2
Gene Cryab
Gene Accession No. P02510, P02511, P23928
Gene Alias 2310001E10Rik; 3526402H21Rik; AACRYA; active transcription factor CREB; alpha B-crystallin; alpha crystallin B; alpha(B)-crystallin; alpha-B-crystallin; Alpha-crystallin B chain; alpha-crystallin B chain-like protein; AV083133; cAMP response element binding protein 1; cAMP responsive element binding protein 1; cAMP-response element-binding protein-1; cAMP-responsive element-binding protein 1; CMD1II; CREB; CREB1; CREB-1; Crya2; Crya-2; CRYAB; CryAB antibody; crystallin alpha B; crystallin, alpha 2; crystallin, alpha B; crystallin, alpha polypeptide 2; CTPP2; CTRCT16; cyclic adenosine 3',5'-monophosphate response element-binding protein CREB; cyclic AMP-responsive element-binding protein 1; epididymis secretory protein Li 101; heat shock protein beta-5; heat shock protein CryAB; heat shock protein HspB5; heat-shock 20 kD like-protein; HEL-S-101; HSPB5; HspB5 protein; MFM2; P23; Renal carcinoma antigen NY-REN-27; rosenthal fiber component; transactivator protein; Y protein
Gene Symbols Cryab
Host Species Mouse
Immunogen Native Alpha B Crystallin.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1410, 25420, 281719
Target Species Human, Rat, Bovine
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.